3-Chloro-6-(methylamino)pyridazine
Supplier: Matrix Scientific
018888-500MG
101807-064EA
280
USD
101807-064
3-Chloro-6-(methylamino)pyridazine
3-Chloro-6-(methylamino)pyridazine
Hazard: Irritant
Formula:
C₅H₆ClN₃ MW: 143.5700 g/mol Boiling Pt: 357 °C Density: 1.338 Flash Pt: 170 °C |
MDL Number:
MFCD02178601 CAS Number: 14959-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...