2-Acetyl-1-naphthol

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-1-hydroxynaphthalene1-Hydroxy-2-acetonaphthone1'-Hydroxy-2'-acetonaphthone1-(1-Hydroxynaphthalen-2-yl)ethan-1-one

018877-500MG
101807-044EA 280 USD
101807-044
2-Acetyl-1-naphthol
2-Acetyl-1-naphthol

Hazard: Irritant

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00003963
CAS Number: 711-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR