3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid
Supplier: Matrix Scientific
018834-500MG
101806-962EA
207.69
USD
101806-962
3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid
3-Amino-3-(1,3-benzodioxol-5-yl)propanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₄ MW: 209.2 g/mol |
MDL Number:
MFCD00736965 CAS Number: 129042-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...