1-(4-Hydroxyphenyl)cyclopentanecarboxylic acid

Supplier: Matrix Scientific

018828-500MG
101806-954EA 400 USD
101806-954
1-(4-Hydroxyphenyl)cyclopentanecarboxylic acid
1-(4-Hydroxyphenyl)cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MDL Number: MFCD00455268
CAS Number: 91496-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR