(3-Formyl-2-methylindol-1-yl)acetic acidethyl ester

Supplier: Matrix Scientific

018801-500MG
101806-902EA 280 USD
101806-902
(3-Formyl-2-methylindol-1-yl)acetic acidethyl ester
(3-Formyl-2-methylindol-1-yl)acetic acid ethyl ester

Hazard: Irritant

Formula: C₁₄H₁₅NO₃
MDL Number: MFCD03083305
CAS Number: 433307-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR