4-Imidazol-1-ylmethylphenylamine

Supplier: Matrix Scientific

Synonyms: 4-[(1H-Imidazol-1-yl)methyl]aniline

018790-500MG
101806-882EA 55.38 USD
101806-882
4-Imidazol-1-ylmethylphenylamine
4-Imidazol-1-ylmethylphenylamine

Hazard: Irritant

Formula: C₁₀H₁₁N₃
MW: 173.2144 g/mol
MDL Number: MFCD06660785
CAS Number: 56643-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR