4-Methoxyindole-2-carboxylic acid

Supplier: Matrix Scientific

018750-1G
101806-802EA 126.15 USD
101806-802
4-Methoxyindole-2-carboxylic acid
4-Methoxyindole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD02664458
CAS Number: 103260-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR