4-Cyclopentyloxy-3-ethoxybenzaldehyde

Supplier: Matrix Scientific

018727-500MG
101806-756EA 280 USD
101806-756
4-Cyclopentyloxy-3-ethoxybenzaldehyde
4-Cyclopentyloxy-3-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD02375349
CAS Number: 524732-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR