2-Chloro-N-(4-piperidin-1-ylmethylphenyl)acetamide hydrochloride

Supplier: Matrix Scientific

018713-500MG
101806-732EA 400 USD
101806-732
2-Chloro-N-(4-piperidin-1-ylmethylphenyl)acetamide hydrochloride
2-Chloro-N-(4-piperidin-1-ylmethylphenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀Cl₂N₂O
MDL Number: MFCD09997563
CAS Number: 1185296-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR