1-(3-Chloro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

018668-500MG
101806-646EA 280 USD
101806-646
1-(3-Chloro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Chloro-4-methylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂ClNO₃
MDL Number: MFCD01429410
CAS Number: 63674-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR