3-(1-Azepanyl)-1-propanamine

Supplier: Matrix Scientific

Synonyms: 3-(Azepan-1-yl)propan-1-amine

018645-500MG
101806-602EA 280 USD
101806-602
3-(1-Azepanyl)-1-propanamine
3-(1-Azepanyl)-1-propanamine

Hazard: Irritant

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
MDL Number: MFCD00020990
CAS Number: 3437-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR