3-[(2-Bromo-4-methylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

022985-500MG
101815-122EA 495.38 USD
101815-122
3-[(2-Bromo-4-methylphenoxy)methyl]piperidine hydrochloride
3-[(2-Bromo-4-methylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO
MDL Number: MFCD09800635
CAS Number: 1185303-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR