3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]piperidine

Supplier: Matrix Scientific

022975-500MG
101815-104EA 315.38 USD
101815-104
3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]piperidine
3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₉H₂₃NO
MDL Number: MFCD08686919
CAS Number: 946713-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR