3-[(Isopentyloxy)methyl]piperidine

Supplier: Matrix Scientific

022964-500MG
101815-086EA 315.38 USD
101815-086
3-[(Isopentyloxy)methyl]piperidine
3-[(Isopentyloxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₁H₂₃NO
MDL Number: MFCD08687949
CAS Number: 946787-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR