3-[(2-Ethoxyethoxy)methyl]piperidine

Supplier: Matrix Scientific

022959-500MG
101815-076EA 315.38 USD
101815-076
3-[(2-Ethoxyethoxy)methyl]piperidine
3-[(2-Ethoxyethoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₀H₂₁NO₂
MDL Number: MFCD08687699
CAS Number: 868067-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR