3-[(2-Methoxyethoxy)methyl]piperidine

Supplier: Matrix Scientific

022958-500MG
101815-074EA 315.38 USD
101815-074
3-[(2-Methoxyethoxy)methyl]piperidine
3-[(2-Methoxyethoxy)methyl]piperidine

Hazard: Irritant

Formula: C₉H₁₉NO₂
MDL Number: MFCD08688038
CAS Number: 868067-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR