N,N-Dimethyl-2-(3-piperidinylmethoxy)-1-ethanamine

Supplier: Matrix Scientific

022957-500MG
101815-072EA 538.46 USD
101815-072
N,N-Dimethyl-2-(3-piperidinylmethoxy)-1-ethanamine
N,N-Dimethyl-2-(3-piperidinylmethoxy)-1-ethanamine

Hazard: Irritant

Formula: C₁₀H₂₂N₂O
MDL Number: MFCD08687683
CAS Number: 946786-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR