5-Bromo-2-[(4-chloro-1-naphthyl)oxy]aniline

Supplier: Matrix Scientific

022951-500MG
101815-060EA 315.38 USD
101815-060
5-Bromo-2-[(4-chloro-1-naphthyl)oxy]aniline
5-Bromo-2-[(4-chloro-1-naphthyl)oxy]aniline

Hazard: Irritant

Formula: C₁₆H₁₁BrClNO
MDL Number: MFCD08686965
CAS Number: 946743-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR