5-Bromo-2-(1-naphthyloxy)aniline

Supplier: Matrix Scientific

022950-500MG
101815-058EA 315.38 USD
101815-058
5-Bromo-2-(1-naphthyloxy)aniline
5-Bromo-2-(1-naphthyloxy)aniline

Hazard: Irritant

Formula: C₁₆H₁₂BrNO
MDL Number: MFCD08687036
CAS Number: 65457-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR