5-Bromo-2-(4-ethylphenoxy)aniline

Supplier: Matrix Scientific

022948-500MG
101815-054EA 303.08 USD
101815-054
5-Bromo-2-(4-ethylphenoxy)aniline
5-Bromo-2-(4-ethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08686994
CAS Number: 946665-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR