5-Bromo-2-(4-isopropylphenoxy)aniline

Supplier: Matrix Scientific

022942-500MG
101815-042EA 315.38 USD
101815-042
5-Bromo-2-(4-isopropylphenoxy)aniline
5-Bromo-2-(4-isopropylphenoxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₆BrNO
MDL Number: MFCD08687011
CAS Number: 946743-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR