5-Bromo-2-(4-chloro-3,5-dimethylphenoxy)aniline

Supplier: Matrix Scientific

022935-500MG
101815-028EA 315.38 USD
101815-028
5-Bromo-2-(4-chloro-3,5-dimethylphenoxy)aniline
5-Bromo-2-(4-chloro-3,5-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₃BrClNO
MDL Number: MFCD08686961
CAS Number: 946665-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR