5-Bromo-2-(3,4-dimethylphenoxy)aniline

Supplier: Matrix Scientific

022929-500MG
101815-016EA 315.38 USD
101815-016
5-Bromo-2-(3,4-dimethylphenoxy)aniline
5-Bromo-2-(3,4-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08686986
CAS Number: 946743-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR