5-Bromo-2-(3-methylphenoxy)aniline

Supplier: Matrix Scientific

022928-500MG
101815-014EA 315.38 USD
101815-014
5-Bromo-2-(3-methylphenoxy)aniline
5-Bromo-2-(3-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₂BrNO
MDL Number: MFCD08687022
CAS Number: 946786-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR