5-Bromo-2-(3-isopropylphenoxy)aniline

Supplier: Matrix Scientific

022927-500MG
101815-012EA 315.38 USD
101815-012
5-Bromo-2-(3-isopropylphenoxy)aniline
5-Bromo-2-(3-isopropylphenoxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₆BrNO
MDL Number: MFCD08687010
CAS Number: 946665-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR