5-Bromo-2-[3-(tert-butyl)phenoxy]aniline

Supplier: Matrix Scientific

022926-500MG
101815-010EA 315.38 USD
101815-010
5-Bromo-2-[3-(tert-butyl)phenoxy]aniline
5-Bromo-2-[3-(tert-butyl)phenoxy]aniline

Hazard: Irritant

Formula: C₁₆H₁₈BrNO
MDL Number: MFCD08686959
CAS Number: 946700-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR