5-Bromo-2-(2-ethylphenoxy)aniline

Supplier: Matrix Scientific

022918-500MG
101814-994EA 315.38 USD
101814-994
5-Bromo-2-(2-ethylphenoxy)aniline
5-Bromo-2-(2-ethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08686993
CAS Number: 946700-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR