5-Bromo-2-(2,4-dimethylphenoxy)aniline

Supplier: Matrix Scientific

022914-500MG
101814-986EA 315.38 USD
101814-986
5-Bromo-2-(2,4-dimethylphenoxy)aniline
5-Bromo-2-(2,4-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08686984
CAS Number: 946700-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR