5-Bromo-2-(2-methoxyphenoxy)aniline

Supplier: Matrix Scientific

022908-500MG
101814-974EA 315.38 USD
101814-974
5-Bromo-2-(2-methoxyphenoxy)aniline
5-Bromo-2-(2-methoxyphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₂BrNO₂
MDL Number: MFCD08687015
CAS Number: 946786-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR