5-Bromo-2-[2-(4-morpholinyl)ethoxy]aniline

Supplier: Matrix Scientific

022898-500MG
101814-954EA 209.23 USD
101814-954
5-Bromo-2-[2-(4-morpholinyl)ethoxy]aniline
5-Bromo-2-[2-(4-morpholinyl)ethoxy]aniline

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂O₂
Melting Pt: 66…67 °C
MDL Number: MFCD08687034
CAS Number: 946786-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR