2-(Benzyloxy)-5-bromoaniline

Supplier: Matrix Scientific

022895-500MG
101814-948EA 315.38 USD
101814-948
2-(Benzyloxy)-5-bromoaniline
2-(Benzyloxy)-5-bromoaniline

Hazard: Irritant

Formula: C₁₃H₁₂BrNO
MDL Number: MFCD08686860
CAS Number: 186797-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR