5-Bromo-2-(3-phenylpropoxy)aniline

Supplier: Matrix Scientific

022890-500MG
101814-938EA 315.38 USD
101814-938
5-Bromo-2-(3-phenylpropoxy)aniline
5-Bromo-2-(3-phenylpropoxy)aniline

Hazard: Irritant

Formula: C₁₅H₁₆BrNO
MDL Number: MFCD08687064
CAS Number: 946699-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR