5-Bromo-2-(phenethyloxy)aniline

Supplier: Matrix Scientific

022886-500MG
101814-930EA 315.38 USD
101814-930
5-Bromo-2-(phenethyloxy)aniline
5-Bromo-2-(phenethyloxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄BrNO
MDL Number: MFCD08687049
CAS Number: 946699-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR