5-Bromo-2-(2-ethoxyethoxy)aniline

Supplier: Matrix Scientific

022884-500MG
101814-926EA 315.38 USD
101814-926
5-Bromo-2-(2-ethoxyethoxy)aniline
5-Bromo-2-(2-ethoxyethoxy)aniline

Hazard: Irritant

Formula: C₁₀H₁₄BrNO₂
MDL Number: MFCD08686989
CAS Number: 946786-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR