5-Bromo-2-[2-(dimethylamino)ethoxy]aniline

Supplier: Matrix Scientific

022881-500MG
101814-920EA 495.38 USD
101814-920
5-Bromo-2-[2-(dimethylamino)ethoxy]aniline
5-Bromo-2-[2-(dimethylamino)ethoxy]aniline

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD08686980
CAS Number: 946743-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR