5-Bromo-2-(2-cyclohexylethoxy)aniline

Supplier: Matrix Scientific

022880-500MG
101814-918EA 315.38 USD
101814-918
5-Bromo-2-(2-cyclohexylethoxy)aniline
5-Bromo-2-(2-cyclohexylethoxy)aniline

Hazard: Irritant

Formula: C₁₄H₂₀BrNO
MDL Number: MFCD08686972
CAS Number: 946786-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR