4-(3-Ethoxyphenoxy)-2-methylaniline

Supplier: Matrix Scientific

022877-500MG
101814-912EA 315.38 USD
101814-912
4-(3-Ethoxyphenoxy)-2-methylaniline
4-(3-Ethoxyphenoxy)-2-methylaniline

Hazard: Irritant

Formula: C₁₅H₁₇NO₂
MDL Number: MFCD08687732
CAS Number: 946743-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR