2-Methyl-4-(2-naphthyloxy)aniline

Supplier: Matrix Scientific

022873-500MG
101814-904EA 315.38 USD
101814-904
2-Methyl-4-(2-naphthyloxy)aniline
2-Methyl-4-(2-naphthyloxy)aniline

Hazard: Irritant

Formula: C₁₇H₁₅NO
MDL Number: MFCD08688288
CAS Number: 946699-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR