Methyl-2-[4-(4-amino-3-methylphenoxy)phenyl]acetate

Supplier: Matrix Scientific

022868-500MG
101814-894EA 315.38 USD
101814-894
Methyl-2-[4-(4-amino-3-methylphenoxy)phenyl]acetate
Methyl-2-[4-(4-amino-3-methylphenoxy)phenyl]acetate

Hazard: Irritant

Formula: C₁₆H₁₇NO₃
MDL Number: MFCD08688145
CAS Number: 946664-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR