2-Methyl-4-(4-methylphenoxy)aniline

Supplier: Matrix Scientific

022865-500MG
101814-888EA 315.38 USD
101814-888
2-Methyl-4-(4-methylphenoxy)aniline
2-Methyl-4-(4-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₅NO
Melting Pt: 54 °C
MDL Number: MFCD08688265
CAS Number: 946699-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR