4-([1,1'-Biphenyl]-4-yloxy)-2-methylphenylamine

Supplier: Matrix Scientific

022859-500MG
101814-876EA 315.38 USD
101814-876
4-([1,1'-Biphenyl]-4-yloxy)-2-methylphenylamine
4-([1,1'-Biphenyl]-4-yloxy)-2-methylphenylamine

Hazard: Irritant

Formula: C₁₉H₁₇NO
MDL Number: MFCD08686946
CAS Number: 946743-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR