4-(2-Chlorophenoxy)-2-methylaniline

Supplier: Matrix Scientific

022827-500MG
101814-812EA 315.38 USD
101814-812
4-(2-Chlorophenoxy)-2-methylaniline
4-(2-Chlorophenoxy)-2-methylaniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD08687436
CAS Number: 946664-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR