2-Methyl-4-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-((tetrahydro-2H-pyran-2-yl)methoxy)aniline

022821-500MG
101814-800EA 315.38 USD
101814-800
2-Methyl-4-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine
2-Methyl-4-(tetrahydro-2H-pyran-2-ylmethoxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MW: 221.3 g/mol
MDL Number: MFCD08688402
CAS Number: 946699-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR