2-Methyl-4-propoxyaniline

Supplier: Matrix Scientific

022808-500MG
101814-772EA 495.38 USD
101814-772
2-Methyl-4-propoxyaniline
2-Methyl-4-propoxyaniline

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD08688374
CAS Number: 857007-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR