4-(Isopentyloxy)-2-methylaniline

Supplier: Matrix Scientific

022807-500MG
101814-770EA 315.38 USD
101814-770
4-(Isopentyloxy)-2-methylaniline
4-(Isopentyloxy)-2-methylaniline

Hazard: Irritant

Formula: C₁₂H₁₉NO
MDL Number: MFCD08687947
CAS Number: 946785-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR