4-(2,3-Dihydro-1H-inden-5-yloxy)-3-fluorophenylamine

Supplier: Matrix Scientific

022790-500MG
101814-736EA 315.38 USD
101814-736
4-(2,3-Dihydro-1H-inden-5-yloxy)-3-fluorophenylamine
4-(2,3-Dihydro-1H-inden-5-yloxy)-3-fluorophenylamine

Hazard: Irritant

Formula: C₁₅H₁₄FNO
MDL Number: MFCD08687588
CAS Number: 937598-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR