3-Fluoro-4-(4-methoxyphenoxy)phenylamine

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-(4-methoxyphenoxy)aniline

022777-500MG
101814-710EA 315.38 USD
101814-710
3-Fluoro-4-(4-methoxyphenoxy)phenylamine
3-Fluoro-4-(4-methoxyphenoxy)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₂FNO₂
MW: 233.24 g/mol
MDL Number: MFCD08687845
CAS Number: 87294-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR