4-(4-Chloro-3,5-dimethylphenoxy)-3-fluoroaniline

Supplier: Matrix Scientific

022776-500MG
101814-708EA 315.38 USD
101814-708
4-(4-Chloro-3,5-dimethylphenoxy)-3-fluoroaniline
4-(4-Chloro-3,5-dimethylphenoxy)-3-fluoroaniline

Hazard: Irritant

Formula: C₁₄H₁₃ClFNO
MDL Number: MFCD08687277
CAS Number: 946742-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR