N-[3-(4-Amino-2-fluorophenoxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

022763-500MG
101814-682EA 315.38 USD
101814-682
N-[3-(4-Amino-2-fluorophenoxy)phenyl]acetamide hydrochloride
N-[3-(4-Amino-2-fluorophenoxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₄ClFN₂O₂
MDL Number: MFCD11506380
CAS Number: 1185094-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR