3-Fluoro-4-(3-fluorophenoxy)aniline

Supplier: Matrix Scientific

022760-500MG
101814-676EA 315.38 USD
101814-676
3-Fluoro-4-(3-fluorophenoxy)aniline
3-Fluoro-4-(3-fluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₉F₂NO
MDL Number: MFCD08687815
CAS Number: 937597-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR